Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
1377.00 EUR - 2568.27 EUR
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
16594560
|
Thermo Scientific™
840-323100 |
GENESYS Validator | GENESYS 40-180, BioMate 160 Spectrophotometers |
2525.00 EUR
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16665492
|
Thermo Scientific™
840-326100 |
GENESYS 40-180 Performance Verification Kit | GENESYS 40-180, BioMate 160 Spectrophotometers |
2000.00 EUR
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15905352
|
Thermo Scientific™
840-293000 |
Validator 2 for INSIGHT – A4 Version | Evolution 200 series and Evolution 350 Spectrophotometers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11947471
|
Thermo Scientific™
699-119300 |
Validator Lite for Evolution 60S, BioMate 3S, GENESYS 10S UV-Vis and GENESYS 10S UV-Vis Bio | Evolution 60S/60; BioMate 3S; GENESYS 10S UV-Vis Bio/10S UV-Vis |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Packages provide:- Information on the validation process
- Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
- Information on pharmacopoeia recommended tests for specific instrument performance
- Installation and instrument certification