Learn More
UVP Model M-10E Mini UV Transilluminators: Filter Assembly
Accessory for UVP Model M-10E Mini UV Transilluminators
Brand: UVP 38-0150-01
Additional Details : Weight : 2.00000kg
Description
- For use with UVP Model M-10E Mini UV Transilluminators
1International model.
Specifications
Filter Assembly | |
3.93 x 3.93 in. | |
10 x 10 cm |
Replacement filter | |
M-10E mini transilluminator |