Learn More
UVP Camera Filter SYBR Green
Allows visualization of fluorophores from 460-470nm
Brand:  UVP 38-0219-01
Additional Details : Weight : 0.04888kg
Product Code. 12810703
  • 355.00 EUR / Each
Estimated Shipment: 12-08-2024
Description
  • Blocks UV and IR radiation emitted from the transilluminator
Specifications
Specifications
50
For use with chromadoc imaging system
Camera Filter Sybr Green