Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
190.27 EUR valid until 2024-05-31
Use promo code "21724" to get your promotional price.
198.83 EUR valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "21724" to get your promotional price.
198.83 EUR valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Click to view available options
Size:
Small
X-Large
2X-Large
3X-Large
Description
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
Specifications
Material | Polyethylene |
Color | White |
Closure Type | Snaps |
Sterility | Non-sterile |