Learn More
DuPont™ Tyvek™ IsoClean Disposable Shoe Covers
IsoClean overshoes are an ideal balance of protection, durability, and comfort.
127.50 EUR
Specifications
Material | Tyvek |
---|---|
Color | White |
Disposable | Single-use |
Closure Type | Elastic Ankle |
Description
- Fixation ties
- Elastics at ankles and gripper slip-retardant full sole
- Tyvek™ garments and accessories are composed of flash spun high-density polythylene, providing an ideal balance of protection, durability and comfort
- Tyvek™ is permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries
- Suitable for use in cleanroom class ISO 6 and higher
- Not clean laundered, not sterilized
Specifications
Tyvek | |
White | |
Single-use | |
Elastic Ankle |