Learn More
Miele Professional™ Trolley for washer-disinfectors
Trolley MF/3 for loading and unloading washer and disinfectors
Brand:  Miele Professional™ 6392900
Product Code. 15410475
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • For easy handling of the mobile instrument unit
  • With lifting technology operated by foot pressure
  • Equipped with 2 lockable castors
  • Infeed height 751 mm, +/- 100 mm
Specifications
Specifications
Stainless Steel
Transport System
Stainless Steel
Washer-disinfectors, laboratory