missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl borate, 98%, Thermo Scientific Chemicals

Catalog Number p-7024518
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
688-74-4
C12H27BO3
230.155
MFCD00009434
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC
This item is not returnable. View return policy
688-74-4
C12H27BO3
230.155
MFCD00009434
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point <-70°C
Density 0.853
Boiling Point 230°C to 235°C
Flash Point 93°C (199°F)
Refractive Index 1.4095
Quantity 100 mL
Beilstein 1703865
Sensitivity Moisture sensitive
Formula Weight 230.16
Percent Purity 98%
Chemical Name or Material Tri-n-butyl borate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'