Learn More
3M™ Tracer™ Replacement Pod
104.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Color:
Blue
Dark Blue
Description
- Reduces noise levels up to 20 dB SNR in accordance with EN 352-2:2020.
- Patented tri-flange design in pre-moulded material makes them a fit for most ear canals.
- Firm, long stem for easy insertion and removal.
Specifications
Color | Dark Blue |
Certifications/Compliance | CE Approved |
Quantity | 50/Pk. |
Product Type | Earplug |