Learn More
Fisherbrand™ Traceable™ Memory Hygrometer/Thermometer
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
- Minimum/maximum humidity and temperature readings
- Auto-off function
- w/Computer output
- w/Programmable audible alarm
Specifications
75% Calibration Standard | |
Programmable Audible Alarm | |
Used for memory hygrometer/thermometer | |
Calibration instructions |