Learn More
TIMM10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11124-2-AP-150UL
This item is not returnable.
View return policy
Description
The mitochondrial protein encoded by this gene belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane, functioning as intermembrane space chaperones for the highly insoluble carrier proteins.Specifications
TIMM10 | |
Polyclonal | |
Unconjugated | |
TIMM10 | |
TIM10, TIM10A, TIMM10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26519, 30059, 64464 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62072, P62073, P62074 | |
Timm10 | |
TIMM10 Fusion Protein Ag1615 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |