Learn More
TIAR Mouse anti-Human, Unconjugated, Clone: 1E8D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66907-1-IG-150UL
This item is not returnable.
View return policy
Description
The TIAR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIAR. This antibody recognizes human, mouse, rat antigen. The TIAR antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
TIAR | |
Monoclonal | |
940 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1E8D12 | |
Unconjugated | |
150 μL | |
7073 | |
IgG1 |