missing translation for 'onlineSavingsMsg'
Learn More

Thymidine, MP Biomedicals™
BIOPHARMA

Catalog Number p-7083094
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
This item is not returnable. View return policy
50-89-5
C10H14N2O5
242.23
IQFYYKKMVGJFEH-UHFFFAOYNA-N
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
5789
CHEBI:17748
1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
This item is not returnable. View return policy
50-89-5
C10H14N2O5
242.23
IQFYYKKMVGJFEH-UHFFFAOYNA-N
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
5789
CHEBI:17748
1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Used in forming hybrid cell lines where cells are incubated in HAT medium. Thymidine, MP Biomedicals, a cell culture reagent, is a nucleoside composed of deoxyribose (a pentose sugar) joined to the pyrimidine base thymine. It is highly stable and appears

Mainly used in forming hybrid cell lines using HAT (hypoxanthine-aminopterin-thymidine) medium to give rise to monoclonal antibodies. Also used in HT and HAT cocktails for hybridoma fusion, selection and cloning.

Deoxythymidine can be phosphorylated with one, two or three phosphoric acid groups, creating respectively dTMP, dTDP or dTTP (deoxythymidine mono-, di-, or triphosphate). The stability of deoxythymidine under standard temperature and pressure (STP) is very high. Thymidine is listed as a chemical teratogen.

Melting Point 185°C to 191°C
Quantity 1 g
Solubility Information Soluble in water, methanol, hot alcohol, hot acetone, hot ethyl acetate pyridine, glacial acetic acid and HCl (20mg/mL, clear, colorless solution); sparingly soluble in hot chloroform.
Optical Rotation 17.5–19.5° (c=1, water)
Formula Weight 242.2
Percent Purity >99.9%
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'