Learn More
Fisherbrand™ Thin-Walled PCR Tubes
Provide for rapid, even heat transfer for reliable amplifications. Fisherbrand™ Thin-Walled PCR Tubes are manufactured from virgin polypropylene.
Brand: Fisherbrand™
Additional Details : Weight : 0.40590kg
Description
- Available in mixed or natural colors
- Available in individual tubes or strips
- Available with a variety of caps - domed, flat, strip
Specifications
Assorted | |
0.5 mL | |
PCR | |
PCR Tube |
Individual tubes | |
Polypropylene | |
Flat |