Learn More
Thermo Scientific™ Polypropylene Centrifuge Bottles
Click to view available options
Capacity (Metric):
400 mL
750 mL
Description
Polypropylene bottle with cap
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 13 oz., 400mL |
Capacity (Metric) | 400 mL |
Capacity (English) | 13 oz. |
Material | Polypropylene |
For Use With (Equipment) | Centrifuges |