Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel Pipette Tips
A variety of tip styles and configurations
Brand: Thermo Scientific™ 3881
60.56 EUR valid until 2024-05-31
Use promo code "21712" to get your promotional price.
Additional Details : Weight : 2.09000kg
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
- Available in blue or clear color
Specifications
1000 μL | |
Sterile | |
DNase, RNase, DNA, ATP and Endotoxin Free | |
Standard | |
Sterile |
Non-Filtered | |
Clear | |
Rack | |
ART |