missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylphosphonium bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as a pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used as a pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Insoluble in water. |
Formula Weight | 171.03 |
Percent Purity | 98% |
Chemical Name or Material | Tetramethylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'