Learn More
Velp Scientifica™ Test Tube Rack
Velp Scientifica™ Test Tube Rack is a fundamental accessory of the Velp Scientifica™ (OCB) Open Circulating Bath.
Brand: Velp Scientifica™ A00000093
66.50 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
- Completely transparent for a constant visual control
- Supports variety of test tubes sizes
Specifications
Test Tube Rack | |
36 11 mm Diamter Tubes | |
36 |
OCB | |
Polycarbonate |