Learn More
TentaGel™ MB-Br, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
Specifications
Color | White to Yellow |
Physical Form | Powder |
Loading | 0.4 to 0.6mmol/g |
Packaging | Glass bottle |
Particle Size | 140 to 170 Micron |
Quantity | 1 g |
Chemical Name or Material | TentaGel™ MB-Br |