Learn More
Techne™ Replacement Block
Parts and accessories to keep your Techne™ 3 Prime thermal cycler working optimally.
1130.00 EUR - 1885.00 EUR
Specifications
For Use With (Equipment) | Thermal Cycler |
---|---|
Product Type | Replacement Block |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
12648875
|
Techne™
99957-99 |
18 x 0.5 mL |
1185.00 EUR
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12688875
|
Techne™
99958-00 |
48 x 0.2 mL |
1885.00 EUR
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12628875
|
Techne™
93945-37 |
24 x 0.2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12608885
|
Techne™
93945-43 |
30 x 0.5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Thermal Cycler | |
Replacement Block |