Learn More
Sulfuric Acid, 2.5M, Chem Lab™
Product Code. 11948514
Click to view available options
Quantity:
1 L
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Specifications
Appearance Colorless
pH <1
Composition Sulfuric acid 2.5 mol/L
Concentration 245.2g H2SO4/l H2O = 5 N (±0.01/20°C)
Density 1.15g/mL
Odor Odorless
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Sulfuric acid 2.5 mol/l
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid