Learn More
Sulfuric Acid, 0.025M, Chem Lab™
Click to view available options
Quantity:
1 L
Specifications
Specifications
Appearance | Colorless |
pH | <2 |
Composition | Sulfuric acid 0.025 mol/L |
Concentration | 2.453g H2SO4/l H2O = 0.05 N (±0.0004/20°C) |
Density | 1g/mL |
Odor | Odorless |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Soluble in Water |