missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxypyridazine, 99.04%, MP Biomedicals™
Click to view available options
Quantity:
1 g
25 g
Specifications
Quantity | 1 g |
Formula Weight | 280.3 |
Percent Purity | 0.9904 |
Physical Form | Crystalline |
Chemical Name or Material | Sulfamethoxypyridazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'