Learn More
BRAND™ Stoppers for Centrifuge Tube
Polyethylene caps prevent centrifuge tubes from spilling during processing.
6.90 EUR - 25.00 EUR
Specifications
Material | Polyethylene |
---|---|
Product Type | Caps |
For Use With (Equipment) | Polypropylene centrifuge tubes |
Quantity | 100/PK |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10579341
|
BRAND™
115370 |
48mL |
17.00 EUR
Pack of 100 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
10662052
|
BRAND™
115376 |
160mL |
25.00 EUR
Pack of 100 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
10674542
|
BRAND™
115360 |
10mL |
6.90 EUR
Pack of 100 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
10374941
|
BRAND™
115374 |
110mL |
25.00 EUR
Pack of 100 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
- Polyethylene construction
- For use with polypropylene centrifuge tubes
- Available in a variety of sizes
Specifications
Polyethylene | |
Caps | |
Polypropylene centrifuge tubes | |
100/PK |