Learn More
SteriBags™ Plain Top Closure Pouches
For use in porous load pre-vacuum autoclaves as well as gamma sterilisers. SteriBags™ Plain Top Closure Pouches have a double folded bottom seal for extra integrity.
Brand: Vernacare BP06102
Additional Details : Weight : 1.00000kg
Description
- Manufactured from high quality wet strength MG bleached kraft paper
- Double folded bottom seal for extra integrit
- Opening along width
- Conforms to BS 6257
Specifications
Code: D | |
BS 6257 | |
Use in porous load pre-vacuum autoclaves as well as gamma sterilizers | |
250 mm |
Autoclavable | |
Plain | |
1000/Pk. | |
140 mm |