Learn More
RUBIS Long Tail Tweezers
Long Tail Tweezers - Inox Steel, Serrated Handle
POA
Specifications
Tip Style | Straight, Round, Blunt, Serrated |
---|---|
Material | Inox Steel |
Product Type | Long Tail Tweezers |
Quantity (No. per Case) | Each |
Type | Tweezers |
Description
- Long Tail Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Serrated handles
- Serrated tips for better grip
- Blunt tips
- Suitable for cryogenics applications
- Strong
Specifications
Straight, Round, Blunt, Serrated | |
Long Tail Tweezers | |
Tweezers |
Inox Steel | |
Each |