Learn More
Bochem™ Stainless Steel Transport Container with Lid and Handle
295.00 EUR - 472.00 EUR
Specifications
Product Type | Transport Container |
---|---|
Material | Stainless Steel (18/10) |
Includes | Lid and Handle |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10199740
|
Bochem™
8307 |
10 L |
472.00 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10463501
|
Bochem™
8300 |
1 L |
295.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10792702
|
Bochem™
8301 |
2 L |
299.00 EUR
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10673701
|
Bochem™
8302 |
3 L |
330.00 EUR
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
10169460
|
Bochem™
8304 |
5 L |
396.00 EUR
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Specifications
Transport Container | |
Stainless Steel (18/10) | |
Lid and Handle |