Learn More
Bochem™ Stainless Steel Mortar
Bochem™ Stainless Steel Mortar for use with pestle.
Brand: Bochem™ 8933
Additional Details : Weight : 0.25000kg
Description
- Stainless Steel
- For use with Stainless Steel pestle
Specifications
1000 mL | |
160mm | |
Stainless Steel (18/10) |