Learn More
Buerkle™ Stainless Steel Conical QualiRod
Conical hand-drill, conically shaped, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.)
80.20 EUR - 130.00 EUR
Product Code | Brand | Length (Metric) Blade | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Blade | Price | Quantity & Availability | |||||
10690543
|
Buerkle™
5311-0001 |
110 mm |
80.20 EUR
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
13597350
|
Buerkle™
5311-0002 |
150 mm |
130.00 EUR
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
A conically shaped hand-drill, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.).- Stainless steel V2A (1.4301)
- Sterilizable
- According to EN/ISO 707
Specifications
Stainless Steel (AISI 304) | |
Autoclavable |