Learn More
Cole-Parmer™ Stuart™ SHM/STAND Homogenizer Stand for Benchtop Applications
Make the most of your homogenizer
Brand: Cole-Parmer™ 04720-09
Additional Details : Weight : 1.00000kg
Description
- Probes available in different sizes and materials
- Stand for benchtop use
- Parts and adapters to expand capabilities
- Compatible with SHM1 and SHM3 Adapts disposable probes to homogenizer
Specifications
Stainless Stand | |
Stainless Steel | |
SHM | |
1 Ea. |