Learn More
Fisherbrand™ Solid PTFE Stirring Rod
Chemically resistant, inert and leak-free
Brand:  Fisherbrand™
Additional Details : Weight : 0.02500kg
Product Code. 16255861
  • 29.85 EUR / Pack of 5
Estimated Shipment: 24-05-2024
Description
  • Chemically resistant, inert and leak-free
  • Use up to 280°C (540°F)
Specifications
Specifications
8 mm
Use up to 280°C (540°F)
PTFE
300 mm