Learn More
Fisherbrand™ Solid PTFE Stirring Rod
Brand:  Fisherbrand™
Additional Details : Weight : 0.02000kg
Product Code. 16245861
  • 26.15 EUR / Pack of 5
Estimated Shipment: 28-06-2024
Specifications
8 mm
Solid PTFE
2/Pk.
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
25g
Mixing of various chemicals
Yes
250 mm