missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼110°C (decomposition) |
Quantity | 5 g |
UN Number | UN3396 |
Beilstein | 4047608 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride. |
Formula Weight | 211.94 |
Percent Purity | 95% |
Chemical Name or Material | Sodium triacetoxyborohydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'