Learn More
Epredia™ Single Cytofunnel™ with White Filter Cards
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
186.00 EUR - 898.00 EUR
Specifications
Color | White |
---|---|
Disposable | Single-use |
Type | Single Cytofunnel™, With Filter Cards and Caps |
Volume (Metric) | 0.1 to 0.5 mL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13127963
|
Epredia™
1102548 |
500/Cs. |
898.00 EUR
Pack of 500 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
12076679
|
Epredia™
5991040 |
50 Pk. |
186.00 EUR
Pack of 50 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- Disposable sample chambers with white filter cards and caps
- Pre-attached filter card
Specifications
White | |
Single-use | |
Single Cytofunnel™, With Filter Cards and Caps | |
0.1 to 0.5 mL |