Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand: BRAND™ 27011
Additional Details : Weight : 2.86000kg
Description
Specifications
To Deliver | |
B | |
Upper end cotton plug | |
Orange | |
360 mm |
DIN EN ISO 835 | |
0.1 mL | |
12 | |
10 mL |