Learn More
Epredia™ EZ Single Cytofunnel™ with Brown Filter Cards
Use the Epredia™ EZ Single Cytofunnel™ with Brown Filter Cards for sample volumes of 0.4 mL or less.
Brand:  Epredia™ A78710004
Additional Details : Weight : 0.50000kg
Product Code. 10006318
  • 88.15 EUR / Pack of 40
Estimated Shipment: 21-05-2024
Description
  • Cell deposition area of 6 mm in diameter (28 mm2)
  • Pre-attached brown filter card for slower absorbency
Specifications
Specifications
Cytofunnel with brown filter cards and caps
40/Cs.
Cytofunnel with Brown Filter Card
0.4 mL