Learn More
TPP Techno Plastic Products™ Serological Pipets
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
60.75 EUR - 138.86 EUR
Product Code | Brand | Volume (Metric) | Graduations | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Color | Price | Quantity & Availability | |||
11371784
|
TPP Techno Plastic Products™
94550 |
50 mL | 1/10 | Purple |
138.86 EUR
Pack of 50 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
11311784
|
TPP Techno Plastic Products™
94005 |
5 mL | 1/10 | Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11341784
|
TPP Techno Plastic Products™
94525 |
25 mL | 2/10 | Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11361784
|
TPP Techno Plastic Products™
94525 |
25 mL | 2/10 | Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11321784
|
TPP Techno Plastic Products™
94010 |
10 mL | 1/10 | Orange |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11391774
|
TPP Techno Plastic Products™
94002 |
2 mL | 1/100 | Green |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
- Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
- Smooth interiors, prevent cell hang-up or clumping inside the pipette
- Excellent optical transparency
- Color coded cotton plugs and dispenser boxes for easy identification
- Convenient, stackable TPP dispenser box
- Single wrapped in lint-free paper/plastic blister
- Slender and short form enables ergonomic working conditions in the sterile hood
- Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
- Highly drop free pipet tip design
- Optimized form of the mouthpiece preserves the rubber of the pipet aid
- Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Specifications
Polystyrene | |
Yes |