Learn More
Cytiva (formerly Pall Lab) Sentino™ Pump Magnetic Funnel Adapter
Sentino pump replacement tubing adapter for magnetic funnels.
Brand:  Cytiva (formerly Pall Lab) 4283
Additional Details : Weight : 0.19000kg
Product Code. 17104301
  • 65.20 EUR / Each
Estimated Shipment: 16-07-2024
Description
  • Designed for use with our Sentino pump
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Polyphenylsulfone