Learn More
Sampling Systems™ Sack Clip
Quick, simple and effective solution to closing sacks. Sampling Systems™ Sack Clip Ssolves the problems of sealing sacks after sampling or dispensing.
Brand:  Sampling Systems™ 8010A-1
Product Code. 15879391
  • 83.95 EUR / Pack of 10
Estimated Shipment: 03-12-2024
Description
  • Quick and easy to apply and remove
  • Ratchet mechanism and secondary positive lock for double security
  • Robust
  • Reusable
  • Made from Polyamide (PA)
  • Simply close sack between the jaws of the Sack Clip
Specifications
Specifications
Polyamide
100 mm jaw length
White
250 mm