Learn More
Epredia™ Richard-Allan Scientific™ Modified Wright Stain Pack
Achieve optimum performance on Hema-Tek™ Slide Stainers with the Epredia™ Richard-Allan Scientific™ Stain Pack.
222.00 EUR
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10539408
|
Epredia™
3000 |
Modified Wright Stain Pack | |||||||
Description
This system consists of a modified polychrome methylene blue-eosin stain (similar to Wright or Giemsa) and based on the original stain proposed by Romanowsky*.Specifications
5 Pk. |