Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Brand: Epredia™ 41754
51.30 EUR valid until 2024-10-31
Use promo code "21839" to get your promotional price.
This item is not returnable.
View return policy
Description
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
- Easy to transport trays ideal for operating rooms and hospitals
- Clear polystyrene containers have screw-top lids
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
- Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Easy to transport trays
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
- Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Stack pack reduces excess packaging
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Dry Specimen Container | |
Polyethylene | |
5.09 L | |
Opaque |
Yes | |
Non-sterile | |
Snap Cap | |
10/Cs. |