Learn More
Trajan Scientific and Medical™ Replacement Plunger for Syringes with Valves
Replacement plungers for gas tight syringes.
Brand: Trajan Scientific and Medical™ 0312176
This item is not returnable.
View return policy
Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
104 mm | |
Replacement Plunger for Syringes with Valves | |
100 mL |