Learn More
RDM1 Mouse anti-Human, Unconjugated, Clone: 1C10B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66941-1-IG-20UL
This item is not returnable.
View return policy
Description
The RDM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RDM1. This antibody recognizes human, mouse, rat antigen. The RDM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
RDM1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1C10B4 | |
Unconjugated | |
20 μL | |
201299 | |
IgG2a |