missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBMX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | RBMX2 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898014
|
Proteintech
17994-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888014
|
Proteintech
17994-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
RBMX2 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
B0BN49, Q8R0F5, Q9Y388 | |
209003, 367930, 51634 | |
RBMX2 Fusion Protein Ag12439 | |
Primary | |
-20°C | |
RBMX2 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CGI 79, RBMX2 | |
Rbmx2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |