Learn More
Reagecon™ Ratio Turbidity Standard
123.00 EUR - 192.00 EUR
Specifications
Type | Ratio Turbidity Standard |
---|---|
For Use With (Equipment) | Ratio Instruments |
Accuracy | ±1% |
Certifications/Compliance | NIST, US EPA Approved |
Product Code | Brand | Volume (Metric) | Turbidity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Turbidity | Price | Quantity & Availability | ||||
11536583
|
Reagecon™
CRSR-50-100 |
100 mL | 50 NTU |
125.00 EUR
100mL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11788455
|
Reagecon™
CRSR-20-100 |
100 mL | 20 NTU |
125.00 EUR
100mL |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
11449159
|
Reagecon™
CRSR-0.2-500 |
500 mL | 0.2 NTU |
192.00 EUR
500mL |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
11556583
|
Reagecon™
CRSR-90-100 |
100 mL | 90 NTU |
125.00 EUR
100mL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11429169
|
Reagecon™
CRSR-2-500 |
500 mL | 2 NTU |
123.00 EUR
500mL |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
Specifications
Ratio Turbidity Standard | |
Ratio Instruments | |
±1% | |
NIST, US EPA Approved |