missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Phenyl-1,2-ethanediol, 99%, Thermo Scientific Chemicals

26.20 EUR - 72.60 EUR

Chemical Identifiers

CAS 16355-00-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00064262
InChI Key PWMWNFMRSKOCEY-QMMMGPOBSA-N
Synonym r---1-phenyl-1,2-ethanediol, r-1-phenylethane-1,2-diol, 1r-1-phenylethane-1,2-diol, r---1-phenylethane-1,2-diol, r-1-phenyl-1,2-ethanediol, styrene glycol,-, unii-2lzm4b71kf, r-+-1-phenylethane-1,2-diol, 2lzm4b71kf
PubChem CID 2724621
IUPAC Name (1R)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338465
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10494.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11328465
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10494.03
1 g
72.60 EUR
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16355-00-3
138.166
PWMWNFMRSKOCEY-QMMMGPOBSA-N
2724621
C1=CC=C(C=C1)C(CO)O
C8H10O2
MFCD00064262
r---1-phenyl-1,2-ethanediol, r-1-phenylethane-1,2-diol, 1r-1-phenylethane-1,2-diol, r---1-phenylethane-1,2-diol, r-1-phenyl-1,2-ethanediol, styrene glycol,-, unii-2lzm4b71kf, r-+-1-phenylethane-1,2-diol, 2lzm4b71kf
(1R)-1-phenylethane-1,2-diol

Specifications

16355-00-3
272°C to 274°C
MFCD00064262
r---1-phenyl-1,2-ethanediol, r-1-phenylethane-1,2-diol, 1r-1-phenylethane-1,2-diol, r---1-phenylethane-1,2-diol, r-1-phenyl-1,2-ethanediol, styrene glycol,-, unii-2lzm4b71kf, r-+-1-phenylethane-1,2-diol, 2lzm4b71kf
−39° (c=3 in Ethanol)
(1R)-1-phenylethane-1,2-diol
2724621
99%
67°C to 69°C
C8H10O2
2355547
PWMWNFMRSKOCEY-QMMMGPOBSA-N
C1=CC=C(C=C1)C(CO)O
138.166
138.17
(R)-(-)-Phenyl-1,2-ethanediol
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only