Learn More
Scilabware Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
Brand:  Scilabware MA1/33
58.66 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 12031603
  • 61.75 EUR / Each
Estimated Shipment: 03-07-2024
Description
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
•With two parallel necks
Specifications
Specifications
24/29
Borosilicate Glass
Multiple neck adapter
24/29