Learn More
Quickfit™ Multiple Glass Joint Adapter with Three Necks
Fit three apparatus together leak-tight. The QuickFit™ multiple joint adapter with three necks is the perfect choice.
Brand:  Quickfit MA4/23
91.72 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.10000kg
Product Code. 11312035
  • 96.55 EUR / Each
Estimated Shipment: 13-06-2024
Description
With three necks:
  • Two vertical
  • One at 45°
Specifications
Specifications
24/29
Multiple adapter