Learn More
Sartorius Stedim Biotech™ Quartz Microfiber Filters, Discs, Grade T293
Especially suited for emission monitoring at temperatures of up to 900°C and wherever filters of the highest purity are needed.
Brand: Sartorius Stedim Biotech™ FT-3-1109-013
Additional Details : Weight : 0.20000kg
Description
- Free of glass fibers and binding agents
- Suitable for Air/gas filtration, analytical sample preparation, particle analysis
- Non-sterile, bulk packed
Specifications
85 g/m2 | |
Disc | |
Non-sterile | |
Hydrophilic | |
900°C | |
13 mm |
White | |
Quartz Microfiber | |
T 293 | |
100/Pk. | |
0.43 mm |