Learn More
Sartorius Stedim Biotech™ Quartz Microfiber Filters, Discs, Grade T293
Especially suited for emission monitoring at temperatures of up to 900°C and wherever filters of the highest purity are needed.
Brand:  Sartorius Stedim Biotech™ FT-3-1109-070
Additional Details : Weight : 0.04200kg
 View more versions of this product
Product Code. 15902880
  • 290.00 EUR / Pack of 50
Estimated Shipment: 09-07-2024
Description
  • Free of glass fibers and binding agents
  • Suitable for Air/gas filtration, analytical sample preparation, particle analysis
  • Non-sterile, bulk packed
Specifications
Specifications
85 g/m2
Disc
Non-sterile
Hydrophilic
900°C
70 mm
White
Quartz Microfiber
T 293
50/Pk.
0.43 mm