Learn More
Agilent BioTek QC Check Solution No. 1, Yellow
QC Check Solution No. 1 is used for absorbance qualification tests
Brand: Agilent BioTek 7120779
71.83 EUR valid until 2024-07-31
Use promo code "21860" to get your promotional price.
Additional Details : Weight : 0.25000kg
Specifications
QC Solution | |
Absorbance Qualification Testing | |
25 mL |